accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74043-83-7

IUPAC Name: 7,12,22,27-tetraazapentacyclo[26.2.2.2³,⁶.2¹³,¹⁶.2¹⁸,²¹]octatriaconta-1(30),3,5,13,15,18,20,28,31,33,35,37-dodecaene
Synonyms: 7,12,22,27-tetraazapentacyclo[26.2.2.2³,⁶.2¹³,¹⁶.2¹⁸,²¹]octatriaconta-1(30),3,5,13,15,18,20,28,31,33,35,37-dodecaene
Molecular Weight (g/mol): 504.72
Molecular Formula: C34H40N4
InChi Key: MSPWJYMWDZWVPW-UHFFFAOYSA-N
SMILES: C1CCNC2=CC=C(CC3=CC=C(NCCCCNC4=CC=C(CC5=CC=C(NC1)C=C5)C=C4)C=C3)C=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results