Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6,20,25-Tetraaza[6.1.6.1]paracyclophane 98.0+%, TCI America™
SDP

Catalog No. T1378100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1378100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T1378100MG Supplier TCI America Supplier No. T1378100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 74043-83-7
Molecular Formula C34H40N4
Molecular Weight (g/mol) 504.72
MDL Number MFCD00142567
InChI Key MSPWJYMWDZWVPW-UHFFFAOYSA-N
PubChem CID 5231738
IUPAC Name 7,12,22,27-tetraazapentacyclo[26.2.2.2³,⁶.2¹³,¹⁶.2¹⁸,²¹]octatriaconta-1(30),3,5,13,15,18,20,28,31,33,35,37-dodecaene
SMILES C1CCNC2=CC=C(CC3=CC=C(NCCCCNC4=CC=C(CC5=CC=C(NC1)C=C5)C=C4)C=C3)C=C2

Specifications

Melting Point 183°C
Color White
Quantity 100 mg
Formula Weight 504.72
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,6,20,25-Tetraaza[6.1.6.1]paracyclophane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.