accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONOOONOO

CAS RN 74124-79-1

IUPAC Name: bis(2,5-dioxopyrrolidin-1-yl) carbonate
Synonyms: bis(2,5-dioxopyrrolidin-1-yl) carbonate
Molecular Weight (g/mol): 256.17
Molecular Formula: C9H8N2O7
InChi Key: PFYXSUNOLOJMDX-UHFFFAOYSA-N
SMILES: O=C(ON1C(=O)CCC1=O)ON1C(=O)CCC1=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (5)
Quantity 
  • (2)
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (4)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (4)
  • (3)