accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CS(R)HONH3CO(R)(S)HO(S)NOSCH3

CAS RN 74149-38-5

IUPAC Name: (3R,5aS,6S,10aR)-6-hydroxy-3-(hydroxymethyl)-2-methyl-3,10a-bis(methylsulfanyl)-1H,2H,3H,4H,5aH,6H,10H,10aH-pyrazino[1,2-a]indole-1,4-dione
Synonyms: (3R,5aS,6S,10aR)-6-hydroxy-3-(hydroxymethyl)-2-methyl-3,10a-bis(methylsulfanyl)-5aH,6H,10H-pyrazino[1,2-a]indole-1,4-dione
Molecular Weight (g/mol): 356.46
Molecular Formula: C15H20N2O4S2
InChi Key: OVBAGMZLGLXSBN-UOVKNHIHSA-N
SMILES: CS[C@@]12CC3=CC=C[C@H](O)[C@H]3N1C(=O)[C@@](CO)(SC)N(C)C2=O
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results