accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH2NNOHOOONa

CAS RN 7415-22-7

IUPAC Name: sodium 2-[(carbamoylmethyl)(carboxymethyl)amino]acetate
Synonyms: sodium 2-[(carbamoylmethyl)(carboxymethyl)amino]acetate
Molecular Weight (g/mol): 212.14
Molecular Formula: C6H9N2NaO5
InChi Key: QCGKKVWCABJQQI-UHFFFAOYSA-M
SMILES: [Na+].NC(=O)CN(CC(O)=O)CC([O-])=O
Quantity 
  • (3)
  • (2)
pH 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)