accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO(S)H2N

CAS RN 74190-66-2

IUPAC Name: (2S)-2-(4-methoxyphenyl)pyrrolidin-1-ium
Synonyms: (2S)-2-(4-methoxyphenyl)pyrrolidin-1-ium
Molecular Weight (g/mol): 178.25
Molecular Formula: C11H16NO
InChi Key: HCBLZPKEGFYHDN-NSHDSACASA-O
SMILES: COC1=CC=C(C=C1)[C@@H]1CCC[NH2+]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results