Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(4-Methoxyphenyl)pyrrolidine 98.0+%, TCI America™
SDP

Catalog No. M24461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24461G Supplier TCI America Supplier No. M24461G

missing translation for 'validMainframeMsg'

Specifications

CAS 74190-66-2
Color Yellow
Boiling Point 112°C
Molecular Formula C11H16NO
Quantity 1 g
MDL Number MFCD01631836
Synonym 4-(2-Pyrrolidinyl)anisole
InChI Key HCBLZPKEGFYHDN-NSHDSACASA-O
SMILES COC1=CC=C(C=C1)[C@@H]1CCC[NH2+]1
IUPAC Name (2S)-2-(4-methoxyphenyl)pyrrolidin-1-ium
Molecular Weight (g/mol) 178.25
PubChem CID 3756957
Formula Weight 177.25
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-(4-Methoxyphenyl)pyrrolidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.