accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74209-34-0

IUPAC Name: 3-bromo-7-nitro-2H-indazole
Synonyms: 3-bromo-7-nitro-2H-indazole
Molecular Weight (g/mol): 242.03
Molecular Formula: C7H4BrN3O2
InChi Key: NFSTZPMYAZRZPC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC2=C(Br)NN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results