CAS RN 7421-93-4
IUPAC Name:
3,4,5,6,6,7-hexachloropentacyclo[6.3.0.0²,⁴.0³,⁷.0⁵,⁹]undecane-10-carbaldehyde
Synonyms:
3,4,5,6,6,7-hexachloropentacyclo[6.3.0.0²,⁴.0³,⁷.0⁵,⁹]undecane-10-carbaldehyde
Molecular Weight (g/mol):
380.9
Molecular Formula:
C12H8Cl6O
InChi Key:
HCTWZIFNBBCVGM-UHFFFAOYNA-N
SMILES:
ClC12C3C4CC(C=O)C5C4C(Cl)(C13Cl)C(Cl)(Cl)C25Cl
1
–
1
of
1
results
1
Endrin Aldehyde, SPEX CertiPrep™
CAS: 7421-93-4 Molecular Formula: C12H8Cl6O Molecular Weight (g/mol): 380.895 InChI Key: HCTWZIFNBBCVGM-YJTREVOLSA-N PubChem CID: 12147604 SMILES: C1C(C2C3C1C4C5(C2(C(C3(C45Cl)Cl)(Cl)Cl)Cl)Cl)C=O