accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 742103-27-1

IUPAC Name: di-tert-butyl(1-methyl-2,2-diphenylcyclopropyl)phosphane
Synonyms: di-tert-butyl(1-methyl-2,2-diphenylcyclopropyl)phosphane
Molecular Weight (g/mol): 352.5
Molecular Formula: C24H33P
InChi Key: QMLPJDVGNRHGJQ-UHFFFAOYNA-N
SMILES: CC(C)(C)P(C(C)(C)C)C1(C)CC1(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results