Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cBRIDP(regR) 97.0+%, TCI America™
SDP

Catalog No. C29791G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29791G Supplier TCI America Supplier No. C29791G

Specifications

CAS 742103-27-1
Melting Point 91°C
Color White-Yellow
Molecular Formula C24H33P
Quantity 1 g
MDL Number MFCD09753012
Synonym Di-tert-butyl(1-methyl-2,2-diphenylcyclopropyl)phosphine
InChI Key QMLPJDVGNRHGJQ-UHFFFAOYSA-N
SMILES CC1(CC1(C2=CC=CC=C2)C3=CC=CC=C3)P(C(C)(C)C)C(C)(C)C
IUPAC Name ditert-butyl-(1-methyl-2,2-diphenylcyclopropyl)phosphane
Molecular Weight (g/mol) 352.502
PubChem CID 21909131
Formula Weight 352.50
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material cBRIDP(regR)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.