accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOSOOHNOHNNH3CCH3N

CAS RN 74222-97-2

IUPAC Name: methyl 2-({[(4,6-dimethylpyrimidin-2-yl)carbamoyl]amino}sulfonyl)benzoate
Synonyms: sulfometuron methyl
Molecular Weight (g/mol): 364.38
Molecular Formula: C15H16N4O5S
InChi Key: ZDXMLEQEMNLCQG-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC1=NC(C)=CC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results