accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHI

CAS RN 7425-27-6

IUPAC Name: 4-iodobutanoic acid
Synonyms: 4-iodobutanoic acid
Molecular Weight (g/mol): 214
Molecular Formula: C4H7IO2
InChi Key: HALXOXQZONRCAB-UHFFFAOYSA-N
SMILES: OC(=O)CCCI

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results