accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74289-57-9

IUPAC Name: chlorobis(3,5-dimethylphenyl)phosphane
Synonyms: chlorobis(3,5-dimethylphenyl)phosphane
Molecular Weight (g/mol): 276.74
Molecular Formula: C16H18ClP
InChi Key: FCEBDAANWYNQMO-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(C)=C1)P(Cl)C1=CC(C)=CC(C)=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)