Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobis(3,5-dimethylphenyl)phosphine, tech. 90%

Catalog No. AAH2588506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2588506 5 g
AAH2588503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2588506 Supplier Thermo Scientific Chemicals Supplier No. H2588506
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 74289-57-9 | C16H18ClP | 276.744 g/mol

Chlorobis(3,5-dimethylphenyl)phosphine is used in the synthesis of (S)- and (R)-2,2?-Bis[bis(3,5-dimethylphenyl)phosphinoyl]-5,5?,6,6?,7,7?,8,8?-octahydro-1,1?-binaphthyl (Xyl-H 8 -BINAPO).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorobis(3,5-dimethylphenyl)phosphine is used in the synthesis of (S)- and (R)-2,2′-Bis[bis(3,5-dimethylphenyl)phosphinoyl]-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-binaphthyl (Xyl-H 8 -BINAPO).

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74289-57-9
Molecular Formula C16H18ClP
Molecular Weight (g/mol) 276.744
MDL Number MFCD01630841
InChI Key FCEBDAANWYNQMO-UHFFFAOYSA-N
Synonym bis 3,5-dimethylphenyl chlorophosphine, chlorobis 3,5-dimethylphenyl phosphine, chlorobis 3,5-dimethylphenyl phosphane, chloro-bis 3,5-dimethylphenyl phosphane, bis 3,5-dimethylphenyl phosphinous chloride, phosphinous chloride, bis 3,5-dimethylphenyl, bis 3,5-dimethylphenyl chlorophosphine, tech, phosphinous chloride,p,p-bis 3,5-dimethylphenyl
PubChem CID 4187520
IUPAC Name chloro-bis(3,5-dimethylphenyl)phosphane
SMILES CC1=CC(=CC(=C1)P(C2=CC(=CC(=C2)C)C)Cl)C

Specifications

Quantity 5 g
UN Number UN3261
Solubility Information Reacts with water.
Formula Weight 276.75
Percent Purity 90%
Grade Technical
Chemical Name or Material Chlorobis(3,5-dimethylphenyl)phosphine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.