CAS RN 7429-90-5
Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis)
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Aluminum powder, -100+325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Aluminum foil, 0.1mm (0.004in) thick, 99.99% (metals basis)
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC Name: aluminum SMILES: [Al]
Aluminum wire, 2.0mm (0.08in) dia, annealed, Puratronic™, 99.9995% (metals basis)
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Aluminum shot, nominally 9.5mm (0.375in) flattened/spherical shape, 99.99% (metals basis)
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Aluminum slug, 3.175mm (0.125in) dia x 3.175mm (0.125in) length, 99.99% (metals basis)
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC Name: aluminum SMILES: [Al]
Aluminum shot, irregular, 15mm and down, 99.9% (metals basis)
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC Name: aluminium SMILES: [Al]
Aluminum powder, -325 mesh, 99.97% (metals basis), Thermo Scientific Chemicals
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Aluminum wire, 0.18mm (0.007in) dia, Puratronic™, 99.999% (metals basis)
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC Name: aluminum SMILES: [Al]
Aluminum wire, 1.0mm (0.04in) dia, annealed, 99+% (metals basis)
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 SMILES: [Al]
Aluminum wire, 1.5mm (0.059in) dia, annealed, Puratronic™, 99.999% (metals basis)
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC Name: aluminium SMILES: [Al]
Aluminum wire, 0.81mm (0.032in) dia, annealed, Puratronic™, 99.999% (metals basis)
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC Name: aluminum SMILES: [Al]
Aluminum foil, 0.025mm (0.001in) thick, 99.999% (metals basis)
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC Name: aluminum SMILES: [Al]
Aluminum rod, 22mm (0.87in) dia, Puratronic™, 99.9995% (metals basis)
CAS: 7429-90-5 Molecular Formula: Al Molecular Weight (g/mol): 26.98 MDL Number: MFCD00134029 InChI Key: XAGFODPZIPBFFR-UHFFFAOYSA-N Synonym: aluminum,metal,powder,aluminio,metana,adom,alumina fibre,aluminum flake,dust,noral aluminum PubChem CID: 5359268 ChEBI: CHEBI:33629 IUPAC Name: aluminum SMILES: [Al]