accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74291-57-9

IUPAC Name: 2-(2,2,2-trifluoroacetyl)-1,2,3,4-tetrahydroisoquinoline-7-sulfonyl chloride
Synonyms: 2-(2,2,2-trifluoroacetyl)-3,4-dihydro-1H-isoquinoline-7-sulfonyl chloride
Molecular Weight (g/mol): 327.7
Molecular Formula: C11H9ClF3NO3S
InChi Key: SXIYEBVAQTUBOQ-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)N1CCC2=C(C1)C=C(C=C2)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results