CAS RN 7430-15-1
IUPAC Name:
N,N,N-triethylanilinium chloride
Synonyms:
N,N,N-triethylanilinium chloride
Molecular Weight (g/mol):
213.75
Molecular Formula:
C12H20ClN
InChi Key:
ICTMDIORIDZWQN-UHFFFAOYSA-M
SMILES:
[Cl-].CC[N+](CC)(CC)C1=CC=CC=C1
1
–
1
of
1
results
1
Triethylphenylammonium Chloride 98.0+%, TCI America™
CAS: 7430-15-1 Molecular Formula: C12H20ClN Molecular Weight (g/mol): 213.75 MDL Number: MFCD00059974 InChI Key: ICTMDIORIDZWQN-UHFFFAOYSA-M Synonym: Phenyltriethylammonium Chloride PubChem CID: 5743346 IUPAC Name: N,N,N-triethylanilinium chloride SMILES: [Cl-].CC[N+](CC)(CC)C1=CC=CC=C1