Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylphenylammonium Chloride 98.0+%, TCI America™
SDP

Catalog No. P024125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P024125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P024125G Supplier TCI America Supplier No. P024125G

Chemical Identifiers

CAS 7430-15-1
Molecular Formula C12H20ClN
Molecular Weight (g/mol) 213.75
MDL Number MFCD00059974
InChI Key ICTMDIORIDZWQN-UHFFFAOYSA-M
Synonym Phenyltriethylammonium Chloride
PubChem CID 5743346
IUPAC Name N,N,N-triethylanilinium chloride
SMILES [Cl-].CC[N+](CC)(CC)C1=CC=CC=C1

Specifications

Color White
Quantity 25 g
Formula Weight 213.75
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Triethylphenylammonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.