accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 7431-45-0

IUPAC Name: 2-phenylpyrimidine
Synonyms: 2-phenylpyrimidine
Molecular Weight (g/mol): 156.19
Molecular Formula: C10H8N2
InChi Key: OXPDQFOKSZYEMJ-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=NC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results