accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7432-21-5

IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid
Synonyms: (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoic acid|Z-TRP-OH
Molecular Weight (g/mol): 338.36
Molecular Formula: C19H18N2O4
InChi Key: AHYFYYVVAXRMKB-KRWDZBQOSA-N
SMILES: OC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)