CAS RN 7439-88-5
Iridium sponge, -100 mesh, 99.95% (metals basis)
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N Synonym: ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Iridium Standard For ICP, 1000μg/mL in 10% HCl, SPEX CertiPrep™
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Iridium, 99.9%, (trace metal basis), powder
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N Synonym: ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Iridium powder, -325 mesh, 99.9% (metals basis)
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N Synonym: ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Iridium, 0.5% on activated carbon powder, reduced, nominally 50% water wet
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N Synonym: ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Iridium powder, -22 mesh, Premion™, 99.99% (metals basis)
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N Synonym: ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Iridium powder, -60 mesh, 99.95% (metals basis)
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N Synonym: ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Iridium wire, 0.25mm (0.01in) dia, 99.8% (metals basis)
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N Synonym: ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Iridium wire, 0.5mm (0.02in) dia, 99.8% (metals basis)
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N Synonym: ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Iridium black, 99.95% (metals basis)
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N Synonym: ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Iridium shot, 6-7mm (0.24-0.28in), 99.95% (metals basis)
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N Synonym: ion,black,metallicum,iridium, elemental,iridium, ion ir4,iridio,hydrido-iridium,atom,powder,sponge PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Iridium, plasma standard solution, Specpure™ Ir 10,000μg/mL
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Iridium Standard For ICP, 1000μg/mL in 10% HCl, Assurance Grade, SPEX CertiPrep™
CAS: 7439-88-5 Molecular Formula: Ir Molecular Weight (g/mol): 192.22 MDL Number: MFCD00011062 InChI Key: GKOZUEZYRPOHIO-UHFFFAOYSA-N PubChem CID: 23924 ChEBI: CHEBI:49666 IUPAC Name: iridium SMILES: [Ir]
Sigma Aldrich Iridium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Iridium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.