Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iridium, plasma standard solution, Specpure™ Ir 10,000μg/mL

Catalog No. AA35752AP
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35752AP 500 mL
AA35752AD 50 mL
AA35752AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35752AP Supplier Thermo Scientific Chemicals Supplier No. 035752AP
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7439-88-5 | Ir | 192.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-88-5
Molecular Formula Ir
Molecular Weight (g/mol) 192.22
MDL Number MFCD00011062
InChI Key GKOZUEZYRPOHIO-UHFFFAOYSA-N
PubChem CID 23924
ChEBI CHEBI:49666
IUPAC Name iridium
SMILES [Ir]

Specifications

Odor Characteristic
Density Approx. 1g/cm3
Physical Form Liquid
Type Analytical Standard
Quantity 500 mL
Grade Specpure
Chemical Name or Material Iridium
Hazard Category H290-H314-H335
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8, Packing Group: II
EINECSNumber 231-595-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.