CAS RN 74392-49-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PPACK II, Trifluoroacetate Salt
Potent and specific irreversible inhibitor of plasma and glandular kallikreins.