Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PPACK II, Trifluoroacetate Salt

Catalog No. 52021910MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-021-910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-021-910MG Supplier MilliporeSigma™ Supplier No. 52021910MG
Add to Cart
Add to Cart

Potent and specific irreversible inhibitor of plasma and glandular kallikreins.

Peptide Sequence: H-D-Phe-Phe-Arg-chloromethylketone, 2TFA
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 95% by HPLC
Target Proteins Plasma and glandular kallikreins
CAS 74392-49-7
Molecular Formula C25H33ClN6O3·2CF3COOH
Formula Weight 729.1g/mol
Solubility H2O
Content And Storage Protect from moisture
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.