CAS RN 7440-16-6
5 Percent Rhodium on Alumina Powder, Reduced, Dry, 70 microns, BASF Catalyst, Spectrum™ Chemical
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N IUPAC Name: rhodium SMILES: [Rh]
Rhodium on activated carbon, 5% Rh, unreduced, dry
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium black, 99.9% (metals basis)
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium wire, 0.5mm (0.02in) dia, 99.8% (metals basis)
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium sponge, -20 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium, plasma standard solution, Specpure™ Rh 1000μg/mL
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
5 Percent Rhodium on Activated Carbon Powder, Reduced, Water Wet, 27 microns, BASF Catalyst, Spectrum™ Chemical
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N IUPAC Name: rhodium SMILES: [Rh]
Rhodium, 5% on alumina powder, C301099-5
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium 5% on Carbon (wetted with ca. 55% Water), TCI America™
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N Synonym: fume,on carbon,rhodium, elemental,rodio,black,rhodium, metal,unii-dmk383dsac,on alumina,rh/al2o3,dmk383dsac PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Rhodium Standard For ICP, 1000μg/mL in 10% HCl, Assurance Grade, SPEX CertiPrep™
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium Standard For ICP/MS, SPEX CertiPrep™
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Rhodium Standard For ICP, 1000μg/mL in 10% HCl, SPEX CertiPrep™
CAS: 7440-16-6 Molecular Formula: Rh Molecular Weight (g/mol): 102.91 MDL Number: MFCD00011201 MFCD03458392 InChI Key: MHOVAHRLVXNVSD-UHFFFAOYSA-N PubChem CID: 23948 ChEBI: CHEBI:33359 IUPAC Name: rhodium SMILES: [Rh]
Sigma Aldrich Rhodium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.