Learn More
Rhodium, 5% on alumina powder, C301099-5
Description
Rhodium is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Rhodium is a useful reagent for the conversation of nitriles to primary amines and the hydrogenation of aromatic rings, pyridines, aldehydes and sugars. It is also used for the reduction of oximes to amines and for the reduction of nitrobenzene to phenylhydroxylamine using hydrazine hydrate as the hydrogen source.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with organic materials and flammable substances.
Specifications
Specifications
| Color | Gray |
| Physical Form | Powder |
| Quantity | 100 g |
| Odor | Odorless |
| Solubility Information | Insoluble in water. |
| Chemical Name or Material | Rhodium, 5% on alumina powder, C301099-5 |
Safety and Handling
| EINECSNumber | 231-125-0 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.