CAS RN 7440-22-4
Silver wire, 1.5mm (0.06in) dia, Premion™, 99.998% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Silver wire, 0.5mm (0.02in) dia, annealed, 99.9% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Silver Thinfoil, 0.005mm (0.0002in) thick, 99.97% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Silver powder, spherical, APS 1.3-3.2 micron, 99.9% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Silver wool, 99.9+% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Silver powder, APS 4-7 micron, 99.9% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Silver powder, -120 mesh, Premion™, 99.995% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Silver wire, 0.203mm (0.008in) dia, hard, 99.95% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Silver, 99.9%, (trace metal basis), wire, 1.0 mm diam., 1ea = 8.2gr
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Silver, 99.9%, (trace metal basis), wire, 0.25 mm diam., 1ea = 10.4gr
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Silver slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.99% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Silver slug, 6.35mm (0.25in) dia x 12.7mm (0.50in) length, 99.99% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Thermo Scientific™ Silver shot, 1-7mm (0.08-0.12in), Premion™, 99.999% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N IUPAC Name: silver SMILES: [Ag]
Silver shot, 1-3mm (0.04-0.12in), Premion, 99.999% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]
Silver foil, 0.5mm (0.02in) thick, annealed, 99.9% (metals basis)
CAS: 7440-22-4 Molecular Formula: Ag Molecular Weight (g/mol): 107.87 MDL Number: MFCD00003397 InChI Key: BQCADISMDOOEFD-UHFFFAOYSA-N Synonym: argentum,metal,atom,colloidal,silver, colloidal,silver, elemental,algaedyn,amalgum,epinall,silber PubChem CID: 23954 ChEBI: CHEBI:9141 IUPAC Name: silver SMILES: [Ag]