CAS RN 7440-42-8
Boron standard solution, for AAS, 1 mg/ml B in water
CAS: 7440-42-8 | B | 10.81 g/mol
Boron powder, amorphous, APS <1 micron, 93-96%, Mg 3% max, Thermo Scientific Chemicals
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N Synonym: boron, metallic,unii-n9e3x5056q,boron, amorphous,powder, crystalline,boro,boron-elemental,boron, elemental,and compounds,boracium,boranylidyne PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron powder, 98% (metals basis), Thermo Scientific Chemicals
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N Synonym: boron, metallic,unii-n9e3x5056q,boron, amorphous,powder, crystalline,boro,boron-elemental,boron, elemental,and compounds,boracium,boranylidyne PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron pieces, crystalline, 1-30mm (0.039-1.2in), 99.4% (metals basis)
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N Synonym: boron, metallic,unii-n9e3x5056q,boron, amorphous,powder, crystalline,boro,boron-elemental,boron, elemental,and compounds,boracium,boranylidyne PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron powder (96+ atomic % B-11), crystalline, 40-230 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N Synonym: boron, metallic,unii-n9e3x5056q,boron, amorphous,powder, crystalline,boro,boron-elemental,boron, elemental,and compounds,boracium,boranylidyne PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron pieces, crystalline, 20mm (0.8in) & down, 99.5% (metals basis)
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N Synonym: boron, metallic,unii-n9e3x5056q,boron, amorphous,powder, crystalline,boro,boron-elemental,boron, elemental,and compounds,boracium,boranylidyne PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron powder, crystalline, Puratronic™, 99.9999% (metals basis), Thermo Scientific Chemicals
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N Synonym: boron, metallic,unii-n9e3x5056q,boron, amorphous,powder, crystalline,boro,boron-elemental,boron, elemental,and compounds,boracium,boranylidyne PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron Powder, amorphous and crystalline, -325 mesh, 98% (Metals basis), Thermo Scientific Chemicals
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N Synonym: boron, metallic,unii-n9e3x5056q,boron, amorphous,powder, crystalline,boro,boron-elemental,boron, elemental,and compounds,boracium,boranylidyne PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron powder, amorphous, -325 mesh, 90%, Mg nominal 5%, Thermo Scientific Chemicals
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N Synonym: boron, metallic,unii-n9e3x5056q,boron, amorphous,powder, crystalline,boro,boron-elemental,boron, elemental,and compounds,boracium,boranylidyne PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron powder, crystalline, -4+40 mesh, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N Synonym: boron, metallic,unii-n9e3x5056q,boron, amorphous,powder, crystalline,boro,boron-elemental,boron, elemental,and compounds,boracium,boranylidyne PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron Standard For ICP, 1000μg/mL in H2O, SPEX CertiPrep™
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron Standard For ICP, 10,000μg/mL in H2O, SPEX CertiPrep™
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron Standard For ICP/MS, SPEX CertiPrep™
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron 11 Isotope Standard For ICP/MS, 10μg/mL in H20, SPEX CertiPrep™
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]
Boron Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™
CAS: 7440-42-8 Molecular Formula: B Molecular Weight (g/mol): 10.81 MDL Number: MFCD00134034 MFCD00151272 InChI Key: ZOXJGFHDIHLPTG-UHFFFAOYSA-N PubChem CID: 5462311 ChEBI: CHEBI:27560 IUPAC Name: boron SMILES: [B]