Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron powder, 98% (metals basis), Thermo Scientific Chemicals

Catalog No. AA4003618
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4003618 50 g
AA4003603 1 g
AA4003609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4003618 Supplier Thermo Scientific Chemicals Supplier No. 04003618
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-42-8
Molecular Formula B
Molecular Weight (g/mol) 10.81
MDL Number MFCD00134034 MFCD00151272
InChI Key ZOXJGFHDIHLPTG-UHFFFAOYSA-N
Synonym boron, metallic, unii-n9e3x5056q, boron, amorphous, powder, crystalline, boro, boron-elemental, boron, elemental, and compounds, boracium, boranylidyne
PubChem CID 5462311
ChEBI CHEBI:27560
IUPAC Name boron
SMILES [B]

Specifications

Color Brown
Physical Form Crystalline Powder
Quantity 50 g
Odor Odorless
Appearance Brown powder
Assay Percent Range 98% (Metals basis)
Density 2.34g/cm3
Chemical Name or Material Boron
Hazard Category H228-H302-H332-H335: Flammable solid. Harmful if swallowed. Harmful if inhaled. May cause respiratory irritation.
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 231-151-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.