CAS RN 7440-48-4
Cobalt powder, -22 mesh, Puratronic™, 99.998% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt powder, 1.6 micron, 99.8% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt, plasma standard solution, Specpure™ Co 1000μg/mL
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt powder, -60 mesh, 99.5% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt pieces, 99.9+% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt rod, 12mm (0.47in) dia, Puratronic™, 99.995% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt foil, 0.5mm (0.02in) thick, Puratronic™, 99.995% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt foil, 0.1mm (0.004in) thick, Puratronic™, 99.995% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt sputtering target, 76.2mm (3.0in) dia x 3.18mm (0.125in) thick, 99.95% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt foil, 2.0mm (0.08in) thick, 99.95% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt foil, 0.1mm (0.004in) thick, 99.95% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt lump, typically 1.5cm cubes, vacuum remelted, low oxygen, 99.9% (metals basis), Thermo Scientific Chemicals
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.95% (metals basis), Thermo Scientific™
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]
Cobalt wire, 0.25mm (0.01in) dia, Puratronic™, 99.995% (metals basis)
CAS: 7440-48-4 Molecular Formula: Co Molecular Weight (g/mol): 58.93 MDL Number: MFCD00010935 InChI Key: GUTLYIVDDKVIGB-UHFFFAOYSA-N Synonym: kobalt,cobalto,moncation,co1+,cobaltum,powder,cobalt, ion co1+,hydrido,atom,hydride PubChem CID: 104730 ChEBI: CHEBI:27638 IUPAC Name: cobalt SMILES: [Co]