Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cobalt pieces, 12mm (0.47in) & down, Vacuum Deposition Grade, 99.5% (metals basis excluding Ni), Ni nominal 1%

Catalog No. AA3637530
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3637530 250 g
AA3637518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3637530 Supplier Thermo Scientific Chemicals Supplier No. 03637530
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7440-48-4 | Co | 58.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Color Gray
Quantity 250 g
Odor Odorless
Particle Size 12mm (0.47 in.) & down
Form Pieces
Percent Purity 99.5%
Assay (metals basis excluding Ni), Ni nominal 1%
Chemical Name or Material Cobalt pieces
Hazard Category H317-H334-H350
Hazard Statement GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c
EINECSNumber 231-158-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.