CAS RN 7440-64-4
Ytterbium powder, -40 mesh, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium foil, 0.64mm (0.025in) thick, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium foil, 0.1mm (0.004in) thick, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium foil, 1.0mm (0.04in) thick, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium ingot, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium foil, 0.3mm (0.01in) thick, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium pieces, sublimed, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium powder, -200 mesh, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium sputtering target, 50.8mm dia x 1.59mm thick
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N Synonym: unii-mnq4o4wsi1,hydride,mnq4o4wsi1,foil, 3n,yterbio,ingot,powder,ii ion,chips, 3n,ingot, 3n PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Thermo Scientific™ Ytterbium ingot, 99.9% (REO)
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N IUPAC Name: ytterbium SMILES: [Yb]
Ytterbium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-64-4 Molecular Formula: Yb Molecular Weight (g/mol): 173.05 MDL Number: MFCD00011286 InChI Key: NAWDYIZEMPQZHO-UHFFFAOYSA-N PubChem CID: 23992 ChEBI: CHEBI:33381 IUPAC Name: ytterbium SMILES: [Yb]
Sigma Aldrich Ytterbium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Ytterbium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Ytterbium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Ytterbium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2