Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Ytterbium ingot, 99.9% (REO)

Catalog No. AA47227KF
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA47227KF 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA47227KF Supplier Thermo Scientific™ Supplier No. 47227KF
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7440-64-4 | Yb | 173.05 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-64-4
Molecular Formula Yb
Molecular Weight (g/mol) 173.05
MDL Number MFCD00011286
InChI Key NAWDYIZEMPQZHO-UHFFFAOYSA-N
IUPAC Name ytterbium
SMILES [Yb]

Specifications

Quantity 1 Ea.
Form Solid
ShelfLife 5 Years
EINECSNumber 231-173-2
RTECSNumber ZG1925000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.