accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFOSOOFFF

CAS RN 74427-22-8

IUPAC Name: 2,2-difluoroethyl trifluoromethanesulfonate
Synonyms: 2,2-difluoroethyl trifluoromethanesulfonate
Molecular Weight (g/mol): 214.11
Molecular Formula: C3H3F5O3S
InChi Key: NKULBUOBGILEAR-UHFFFAOYSA-N
SMILES: FC(F)COS(=O)(=O)C(F)(F)F
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results