accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3(S)(R)HO

CAS RN 7443-70-1

IUPAC Name: (1R,2S)-2-methylcyclohexan-1-ol
Synonyms: 2-methylcyclohexanol (cis)
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: NDVWOBYBJYUSMF-NKWVEPMBSA-N
SMILES: C[C@H]1CCCC[C@H]1O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)