Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-2-Methylcyclohexanol 99+%, Thermo Scientific™

Catalog No. AC201280050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC201280050 5 g
AC201280010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC201280050 Supplier Thermo Scientific Chemicals Supplier No. 201280050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7443-70-1
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00064169,MFCD00001443
InChI Key NDVWOBYBJYUSMF-NKWVEPMBSA-N
Synonym cis-2-methylcyclohexanol, 1s,2r-2-methylcyclohexan-1-ol, cyclohexanol, 2-methyl-, 1s,2r, 2alpha-methylcyclohexan-1alpha-ol, 1s,2r-2-methyl-1-cyclohexanol
PubChem CID 5325949
IUPAC Name (1R,2S)-2-methylcyclohexan-1-ol
SMILES C[C@H]1CCCC[C@H]1O

Specifications

Color Colorless
Density 0.936
Infrared Spectrum Authentic
Refractive Index 1.4640 to 1.4660 (20°C, 589nm)
Linear Formula CH3C6H10OH
Quantity 5 g
Beilstein 06, II, 17
Formula Weight 114.19
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material cis-2-Methylcyclohexanol, 99+%
Show More Show Less
EINECSNumber 231-187-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.