accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74472-48-3

IUPAC Name: 2,2',3,4,4',6,6'-heptachloro-1,1'-biphenyl
Synonyms: 2,2',3,4,4',6,6'-heptachloro-1,1'-biphenyl
Molecular Weight (g/mol): 395.31
Molecular Formula: C12H3Cl7
InChi Key: OBIUJJSQKPGKME-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(C(Cl)=C1)C1=C(Cl)C=C(Cl)C(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results