Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2',3,4,4',6,6'-Heptachlorobiphenyl Solution, 100μg/mL in Hexane, Ultra Scientific

Catalog No. USRPC168S
Encompass
Change view
Click to view available options
Quantity:
1 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USRPC168S 1 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. USRPC168S Supplier Agilent Technologies Supplier No. RPC168S
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 74472-48-3 , 110-54-3
Molecular Formula C12H3Cl7
Molecular Weight (g/mol) 395.306
InChI Key OBIUJJSQKPGKME-UHFFFAOYSA-N
PubChem CID 92392
IUPAC Name 1,2,3,5-tetrachloro-4-(2,4,6-trichlorophenyl)benzene
SMILES C1=C(C=C(C(=C1Cl)C2=C(C(=C(C=C2Cl)Cl)Cl)Cl)Cl)Cl

Specifications

Physical Form Liquid
Quantity 1 x 2 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material 2,2',3,4,4',6,6'-Heptachlorobiphenyl Solution, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.