accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7452-59-7

IUPAC Name: octyl carbonochloridate
Synonyms: octyl carbonochloridate
Molecular Weight (g/mol): 192.68
Molecular Formula: C9H17ClO2
InChi Key: VFXVAXFIFHSGNR-UHFFFAOYSA-N
SMILES: CCCCCCCCOC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results