Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octyl Chloroformate 97.0+%, TCI America™
SDP

Catalog No. C072425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C072425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C072425ML Supplier TCI America Supplier No. C072425ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7452-59-7
Molecular Formula C9H17ClO2
Molecular Weight (g/mol) 192.683
MDL Number MFCD00000652
InChI Key VFXVAXFIFHSGNR-UHFFFAOYSA-N
Synonym octyl chloroformate, n-octyl chloroformate, chloroformic acid n-octyl ester, carbonochloridic acid, octyl ester, chloroformic acid octyl ester, n-octylchloroformate, octyl chlorocarboxylate, octyl chloridocarbonate #, acmc-209ov2
PubChem CID 81956
IUPAC Name octyl carbonochloridate
SMILES CCCCCCCCOC(=O)Cl

Specifications

Color Colorless
Boiling Point 123°C
Quantity 25 mL
UN Number 3277
Formula Weight 192.68
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material n-Octyl Chloroformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.