accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiHNSiCH3

CAS RN 7453-26-1

IUPAC Name: bis(methyldiphenylsilyl)amine
Synonyms: bis(methyldiphenylsilyl)amine
Molecular Weight (g/mol): 409.68
Molecular Formula: C26H27NSi2
InChi Key: YFONAHAKNVIHPT-UHFFFAOYSA-N
SMILES: C[Si](N[Si](C)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results