Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethyl-1,1,3,3-tetraphenyldisilazane, 97%, Thermo Scientific™

Catalog No. AC313720010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC313720010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC313720010 Supplier Thermo Scientific Chemicals Supplier No. 313720010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Silylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7453-26-1
Molecular Formula C26H27NSi2
Molecular Weight (g/mol) 409.68
MDL Number MFCD00042831
InChI Key YFONAHAKNVIHPT-UHFFFAOYSA-N
Synonym bis methyldiphenylsilyl amine, 1,3-dimethyl-1,1,3,3-tetraphenyldisilazane, 1,1,3,3-tetraphenyl-1,3-dimethyldisilazane, tpdmds, 1-methyl-n-methyl diphenyl silyl-1,1-diphenylsilanamine, 1,1,3,3-tetraphenyl-1,3-dimethyl-disilazane, 1-methyl-n-methyldiphenylsilyl-1,1-diphenylsilylamine, methyl-methyl diphenyl silyl amino-phenylsilyl benzene, silanamine,1-methyl-n-methyldiphenylsilyl-1,1-diphenyl, 1,3-dimethyl-1,1,3,3-tetraphenyldisilazane nt
PubChem CID 81957
IUPAC Name bis(methyldiphenylsilyl)amine
SMILES C[Si](N[Si](C)(C1=CC=CC=C1)C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula NH(H3CSi(C6H5))
Quantity 1 g
Formula Weight 409.68
Chemical Name or Material 1,3-Dimethyl-1,1,3,3-tetraphenyldisilazane, 97%
EINECSNumber 231-227-5
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.