accessibility menu, dialog, popup

UserName

Viewing profile as user:
FNNOH2NFFONH2F

CAS RN 745833-23-2

IUPAC Name: 2-(2,4-difluorophenyl)-6-[1-(2,6-difluorophenyl)carbamoylamino]pyridine-3-carboxamide
Synonyms: 2-(2,4-difluorophenyl)-6-[1-(2,6-difluorophenyl)carbamoylamino]pyridine-3-carboxamide
Molecular Weight (g/mol): 404.33
Molecular Formula: C19H12F4N4O2
InChi Key: FYSRKRZDBHOFAY-UHFFFAOYSA-N
SMILES: NC(=O)N(C1=NC(=C(C=C1)C(N)=O)C1=C(F)C=C(F)C=C1)C1=C(F)C=CC=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results