Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VX-702 98.0+%, TCI America™
SDP

Catalog No. V014725MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V014725MG 25 mg
V0147100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V014725MG Supplier TCI America Supplier No. V014725MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 745833-23-2 | C19H12F4N4O2 | 404.33 g/mol

Chemical Identifiers

CAS 745833-23-2
Molecular Formula C19H12F4N4O2
Molecular Weight (g/mol) 404.33
MDL Number MFCD11616590
InChI Key FYSRKRZDBHOFAY-UHFFFAOYSA-N
Synonym 6-[(Aminocarbonyl)(2,6-difluorophenyl)amino]-2-(2,4-difluorophenyl)-3-pyridinecarboxamide, 2-(2,4-Difluorophenyl)-6-(1-[2,6-difluorophenyl)ureido]nicotinamide
PubChem CID 10341154
IUPAC Name 2-(2,4-difluorophenyl)-6-[1-(2,6-difluorophenyl)carbamoylamino]pyridine-3-carboxamide
SMILES NC(=O)N(C1=NC(=C(C=C1)C(N)=O)C1=C(F)C=C(F)C=C1)C1=C(F)C=CC=C1F

Specifications

Color White
Quantity 25 mg
Formula Weight 404.32
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material VX-702
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.