accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 7460-84-6

IUPAC Name: (oxiran-2-yl)methyl octadecanoate
Synonyms: oxiran-2-ylmethyl octadecanoate
Molecular Weight (g/mol): 340.55
Molecular Formula: C21H40O3
InChi Key: OUQGOXCIUOCDNN-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC1CO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results