Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycidyl Stearate 96.0+%, TCI America™
SDP

Catalog No. S03665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03665G 5 g
S036625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03665G Supplier TCI America Supplier No. S03665G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7460-84-6
Molecular Formula C21H40O3
Molecular Weight (g/mol) 340.55
MDL Number MFCD00059110
InChI Key OUQGOXCIUOCDNN-UHFFFAOYNA-N
Synonym Stearic Acid Glycidyl Ester
PubChem CID 62642
ChEBI CHEBI:82470
IUPAC Name (oxiran-2-yl)methyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC1CO1

Specifications

Melting Point 53°C
Color White
Boiling Point 193°C
Quantity 5 g
Formula Weight 340.55
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Glycidyl Stearate
RTECSNumber WI3500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.