accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74610-55-2

IUPAC Name: 2,3-dihydroxybutanedioic acid; 2-[6-({5-[(4,5-dihydroxy-4,6-dimethyloxan-2-yl)oxy]-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl}oxy)-16-ethyl-4-hydroxy-15-{[(5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl)oxy]methyl}-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde
Synonyms: (.+-.)-tartaric acid; 2-[6-({5-[(4,5-dihydroxy-4,6-dimethyloxan-2-yl)oxy]-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl}oxy)-16-ethyl-4-hydroxy-15-{[(5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl)oxy]methyl}-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde
Molecular Weight (g/mol): 1066.2
Molecular Formula: C50H83NO23
InChi Key: ICVKYYINQHWDLM-UHFFFAOYNA-N
SMILES: OC(C(O)C(O)=O)C(O)=O.CCC1OC(=O)CC(O)C(C)C(OC2OC(C)C(OC3CC(C)(O)C(O)C(C)O3)C(C2O)N(C)C)C(CC=O)CC(C)C(=O)C=CC(C)=CC1COC1OC(C)C(O)C(OC)C1OC
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Type 
  • (3)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)