Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tylosin tartrate salt, MP Biomedicals™

Catalog No. ICN19345405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19345405 5 g
ICN19345401 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19345405 Supplier MP Biomedicals, Inc Supplier No. 0219345405
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 74610-55-2
Molecular Formula C46H77NO17·C4H6O6

Specifications

Physical Form Powder
Color Yellow
Quantity 5 g
Formula Weight 1066.2
Grade Not Available
Chemical Name or Material Tylosin tartrate salt
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.