accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74711-43-6

IUPAC Name: 2-{10-oxo-2-thiatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-6-yl}propanoic acid
Synonyms: Peon zaltoprofen
Molecular Weight (g/mol): 298.36
Molecular Formula: C17H14O3S
InChi Key: MUXFZBHBYYYLTH-UHFFFAOYNA-N
SMILES: CC(C(O)=O)C1=CC=C2SC3=CC=CC=C3C(=O)CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results